Assay Details
| Gene Symbol: LOC4350125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762416.2 | XP_015617902.1 | 2 - 3 | 272 | 125 | |
| XM_015762417.2 | XP_015617903.1 | 2 - 3 | 289 | 125 | ||
| GenBank mRNA | AK101773.1 | - | 2 - 3 | 264 | 125 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015762416.2, XM_015762417.2 transcripts.
Set Membership: |
|

LOC4350125
Genomic Map