Assay Details
| Gene Symbol: LOC9268663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760019.2 | XP_015615505.1 | 1 - 2 | 3115 | 57 | |
| GenBank mRNA | AK243563.1 | - | 1 - 2 | 2834 | 57 | |
More Information
Set Membership: |
|

LOC9268663
Genomic Map