Assay Details
| Gene Symbol: LOC4350215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762536.2 | XP_015618022.1 | 2 - 3 | 1279 | 69 | |
| XM_015762537.2 | XP_015618023.1 | 3 - 4 | 1130 | 69 | ||
| XM_015762538.2 | XP_015618024.1 | 3 - 4 | 1122 | 69 | ||
| XM_015762539.2 | XP_015618025.1 | 3 - 4 | 1109 | 69 | ||
| GenBank mRNA | AK065745.1 | - | 3 - 4 | 1133 | 69 | |
| AK069670.1 | - | 1 - 2 | 522 | 69 | ||
| AK072808.1 | - | 3 - 4 | 1051 | 69 | ||
More Information
Set Membership: |
|

LOC4350215
Genomic Map