Assay Details
| Gene Symbol: LOC4350215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762536.2 | XP_015618022.1 | 3 - 4 | 1823 | 79 | |
| XM_015762537.2 | XP_015618023.1 | 4 - 5 | 1674 | 79 | ||
| XM_015762538.2 | XP_015618024.1 | 4 - 5 | 1666 | 79 | ||
| XM_015762539.2 | XP_015618025.1 | 4 - 5 | 1653 | 79 | ||
| GenBank mRNA | AK065745.1 | - | 4 - 5 | 1677 | 79 | |
| AK069670.1 | - | 2 - 3 | 1066 | 79 | ||
More Information
Set Membership: |
|

LOC4350215
Genomic Map