Assay Details
| Gene Symbol: LOC4350524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762266.2 | XP_015617752.1 | 10 - 11 | 1430 | 77 | |
| GenBank mRNA | AK065321.1 | - | 10 - 11 | 1430 | 77 | |
More Information
Set Membership: |
|

LOC4350524
Genomic Map