Assay Details
| Gene Symbol: LOC4350609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761031.2 | XP_015616517.1 | 2 - 3 | 1383 | 107 | |
| GenBank mRNA | AK109915.1 | - | 2 - 3 | 1376 | 107 | |
More Information
Set Membership: |
|

LOC4350609
Genomic Map