Assay Details
| Gene Symbol: LOC4328780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768521.2 | XP_015624007.1 | 1 - 2 | 346 | 148 | |
| GenBank mRNA | AK289142.1 | - | 1 - 2 | 207 | 148 | |
More Information
Set Membership: |
|

LOC4328780
Genomic Map