Assay Details
| Gene Symbol: LOC4329563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772216.2 | XP_015627702.1 | 2 - 3 | 265 | 116 | |
| GenBank mRNA | AK101459.1 | - | 2 - 3 | 210 | 116 | |
| Gene Symbol: LOC4324655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK102935.1 | - | 201 | 116 | ||
More Information
Set Membership: |
|

LOC4329563
Genomic Map