Assay Details
| Gene Symbol: LOC4332487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773335.2 | XP_015628821.1 | 1 - 2 | 149 | 141 | |
| GenBank mRNA | AK059587.1 | - | 1 - 2 | 128 | 141 | |
| AK103162.1 | - | 1 - 2 | 310 | 141 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015773335.2 transcripts.
Set Membership: |
|

LOC4332487
Genomic Map