Assay Details
| Gene Symbol: LOC4333014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774889.2 | XP_015630375.1 | 1 - 2 | 703 | 133 | |
| GenBank mRNA | AB122057.1 | - | 1 - 2 | 700 | 133 | |
| AK059247.1 | - | 1 - 2 | 276 | 133 | ||
| AK061085.1 | - | 1 - 2 | 101 | 133 | ||
More Information
Set Membership: |
|

LOC4333014
Genomic Map