Assay Details
| Gene Symbol: LOC4347225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756317.2 | XP_015611803.1 | 1 - 2 | 226 | 106 | |
| GenBank mRNA | AK058296.1 | - | 1 - 2 | 226 | 106 | |
More Information
Set Membership: |
|

LOC4347225
Genomic Map