Assay Details
| Gene Symbol: LOC4352858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763260.2 | 1 | 654 | 129 | ||
| GenBank mRNA | AK243287.1 | - | 1 | 599 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4352858| Gene Symbol: LOC4352858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763260.2 | 1 | 654 | 129 | ||
| GenBank mRNA | AK243287.1 | - | 1 | 599 | 129 | |
Set Membership: |
|