Assay Details
| Gene Symbol: LOC4326903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765850.2 | XP_015621336.1 | 2 | 590 | 133 | |
| GenBank mRNA | AK058646.1 | - | 2 | 555 | 133 | |
| BT129067.1 | - | 2 | 548 | 133 | ||
More Information
Set Membership: |
|

LOC4326903
Genomic Map