Assay Details
| Gene Symbol: LOC9266786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK242866.1 | - | 158 | 153 | ||
| Gene Symbol: LOC4323897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789038.2 | XP_015644524.1 | 2 | 1111 | 153 | |
| GenBank mRNA | AK108259.1 | - | 2 | 957 | 153 | |
| AK288851.1 | - | 2595 | 153 | |||
More Information
Set Membership: |
|

LOC9266786
Genomic Map