Assay Details
| Gene Symbol: LOC4328717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768166.2 | XP_015623652.1 | 7 | 1877 | 85 | |
| XM_015768167.2 | XP_015623653.1 | 7 | 1874 | 85 | ||
| GenBank mRNA | AF513384.1 | - | 7 | 1850 | 85 | |
More Information
Set Membership: |
|

LOC4328717
Genomic Map