Assay Details
| Gene Symbol: LOC4330002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768686.2 | XP_015624172.1 | 2 | 1611 | 151 | |
| GenBank mRNA | AK070657.1 | - | 2 | 1555 | 151 | |
More Information
Set Membership: |
|

LOC4330002
Genomic Map