Assay Details
| Gene Symbol: LOC4331608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775609.2 | XP_015631095.1 | 1 | 166 | 125 | |
| GenBank mRNA | AK058322.1 | - | 1 | 146 | 125 | |
| AK060994.1 | - | 1 | 146 | 125 | ||
| AK068986.1 | - | 1 | 150 | 125 | ||
| AK104123.1 | - | 1 | 148 | 125 | ||
| KM051838.1 | - | 1 | 123 | 125 | ||
| Gene Symbol: LOC4326159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287579.1 | - | 1880 | 125 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775609.2 transcripts.
Set Membership: |
|

LOC4331608
Genomic Map