Assay Details
| Gene Symbol: LOC4332141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775378.2 | XP_015630864.1 | 2 | 3695 | 151 | |
| GenBank mRNA | AK121286.1 | - | 2 | 3252 | 151 | |
More Information
Set Membership: |
|

LOC4332141
Genomic Map