Assay Details
| Gene Symbol: LOC4332166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774308.2 | XP_015629794.1 | 4 | 774 | 145 | |
| GenBank mRNA | AK064054.1 | - | 4 | 649 | 145 | |
| BT129127.1 | - | 4 | 650 | 145 | ||
More Information
Set Membership: |
|

LOC4332166
Genomic Map