Assay Details
| Gene Symbol: LOC4334273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777449.2 | XP_015632935.1 | 2 | 3506 | 129 | |
| GenBank mRNA | AK102670.1 | - | 2 | 2853 | 129 | |
More Information
Set Membership: |
|

LOC4334273
Genomic Map