Assay Details
| Gene Symbol: LOC107277808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776490.2 | XP_015631976.1 | 2 | 1192 | 137 | |
| GenBank mRNA | AK060311.1 | - | 1 | 1042 | 136 | |
| AK061411.1 | - | 2 | 885 | 138 | ||
| AK119609.1 | - | 2 | 946 | 136 | ||
More Information
Set Membership: |
|

LOC107277808
Genomic Map