Assay Details
| Gene Symbol: LOC4336456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778878.2 | XP_015634364.1 | 3 | 1060 | 116 | |
| GenBank mRNA | AK061581.1 | - | 3 | 1049 | 116 | |
More Information
Set Membership: |
|

LOC4336456
Genomic Map