Assay Details
| Gene Symbol: LOC4337399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK068623.1 | - | 2 | 1386 | 160 | |
| AK106166.1 | - | 2 | 1344 | 160 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4337399| Gene Symbol: LOC4337399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK068623.1 | - | 2 | 1386 | 160 | |
| AK106166.1 | - | 2 | 1344 | 160 | ||
Set Membership: |
|