Assay Details
| Gene Symbol: LOC4338944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783899.2 | XP_015639385.1 | 3 | 1832 | 137 | |
| XM_015783900.2 | XP_015639386.1 | 3 | 1829 | 137 | ||
| XM_015783901.2 | XP_015639387.1 | 3 | 1836 | 137 | ||
| XM_015783902.2 | XP_015639388.1 | 3 | 1836 | 137 | ||
| GenBank mRNA | AK068137.1 | - | 3 | 1754 | 137 | |
More Information
Set Membership: |
|

LOC4338944
Genomic Map