Assay Details
| Gene Symbol: LOC4339064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784076.2 | XP_015639562.1 | 1 | 314 | 103 | |
| GenBank mRNA | AK065288.1 | - | 1 | 184 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784076.2 transcripts.
Set Membership: |
|

LOC4339064
Genomic Map