Assay Details
| Gene Symbol: LOC4339548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782389.2 | XP_015637875.1 | 3 | 1068 | 161 | |
| GenBank mRNA | AK108452.1 | - | 3 | 1054 | 161 | |
More Information
Set Membership: |
|

LOC4339548
Genomic Map