Assay Details
| Gene Symbol: LOC4342220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791336.2 | XP_015646822.1 | 1 | 355 | 122 | |
| GenBank mRNA | AK063942.1 | - | 1 | 351 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791336.2 transcripts.
Set Membership: |
|

LOC4342220
Genomic Map