Assay Details
| Gene Symbol: LOC4342580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792264.2 | XP_015647750.1 | 4 | 666 | 107 | |
| GenBank mRNA | AK059157.1 | - | 1 | 82 | 107 | |
More Information
Set Membership: |
|

LOC4342580
Genomic Map