Assay Details
Gene Symbol: LOC4344557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794303.2 | XP_015649789.1 | 2 | 2262 | 108 | |
GenBank mRNA | AK059130.1 | - | 2 | 2249 | 108 | |
AK098961.1 | - | 2 | 2193 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
