Assay Details
| Gene Symbol: LOC4346795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755511.2 | XP_015610997.1 | 2 | 1360 | 99 | |
| XM_015755512.2 | XP_015610998.1 | 3 | 1242 | 99 | ||
| GenBank mRNA | AK072740.1 | - | 2 | 1352 | 99 | |
More Information
Set Membership: |
|

LOC4346795
Genomic Map