Assay Details
| Gene Symbol: LOC4348019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757283.2 | XP_015612769.1 | 4 | 1543 | 139 | |
| XM_015757284.2 | XP_015612770.1 | 3 | 1651 | 139 | ||
| GenBank mRNA | AK102940.1 | - | 3 | 1651 | 139 | |
More Information
Set Membership: |
|

LOC4348019
Genomic Map