Assay Details
| Gene Symbol: LOC4349035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758931.2 | XP_015614417.1 | 2 | 1384 | 145 | |
| GenBank mRNA | AK071551.1 | - | 2 | 1353 | 145 | |
More Information
Set Membership: |
|

LOC4349035
Genomic Map