Assay Details
| Gene Symbol: LOC4351248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765174.2 | XP_015620660.1 | 1 | 549 | 121 | |
| GenBank mRNA | AK068622.1 | - | 1 | 551 | 121 | |
More Information
Set Membership: |
|

LOC4351248
Genomic Map