Assay Details
| Gene Symbol: LOC4351591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763917.2 | XP_015619403.1 | 2 | 1964 | 130 | |
| GenBank mRNA | AK110577.1 | - | 2 | 1643 | 130 | |
More Information
Set Membership: |
|

LOC4351591
Genomic Map