Assay Details
| Gene Symbol: AGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194304.2 | NP_001181233.1 | 665 | 69 | ||
| XM_015133907.2 | XP_014989393.1 | 630 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGR2| Gene Symbol: AGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194304.2 | NP_001181233.1 | 665 | 69 | ||
| XM_015133907.2 | XP_014989393.1 | 630 | 69 | |||
Set Membership: |
|