Assay Details
| Gene Symbol: DDX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115626.2 | XP_014971112.1 | 1619 | 72 | ||
| XM_015115627.2 | XP_014971113.1 | 1767 | 72 | |||
| GenBank mRNA | JV046327.1 | - | 1552 | 72 | ||
| JV643986.1 | - | 1549 | 72 | |||
More Information
Set Membership: |
|

DDX10
Genomic Map