Assay Details
| Gene Symbol: FBXO25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261096.1 | NP_001248025.1 | 734 | 75 | ||
| XM_015144664.2 | XP_015000150.1 | 953 | 75 | |||
| XM_015144669.2 | XP_015000155.1 | 1464 | 75 | |||
| XM_015144670.2 | XP_015000156.1 | 952 | 75 | |||
| XM_015144671.2 | XP_015000157.1 | 461 | 75 | |||
| XM_028852276.1 | XP_028708109.1 | 894 | 75 | |||
| XM_028852277.1 | XP_028708110.1 | 893 | 75 | |||
| XM_028852278.1 | XP_028708111.1 | 952 | 75 | |||
| GenBank mRNA | JU333944.1 | - | 762 | 75 | ||
| JU333945.1 | - | 762 | 75 | |||
| JU475130.1 | - | 734 | 75 | |||
More Information
Set Membership: |
|

FBXO25
Genomic Map