Assay Details
| Gene Symbol: BRMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266716.1 | NP_001253645.1 | 327 | 87 | ||
| XM_015113540.2 | XP_014969026.1 | 417 | 87 | |||
| XM_015113541.2 | XP_014969027.1 | 417 | 87 | |||
| GenBank mRNA | JU321377.1 | - | 333 | 87 | ||
| JU478198.1 | - | 386 | 87 | |||
| JV667219.1 | - | 327 | 87 | |||
More Information
Set Membership: |
|

BRMS1
Genomic Map