Assay Details
| Gene Symbol: PHACTR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132959.2 | XP_014988445.1 | 2037 | 72 | ||
| XM_015132961.1 | XP_014988447.1 | 2037 | 72 | |||
| XM_028837191.1 | XP_028693024.1 | 2774 | 72 | |||
| XM_028837198.1 | XP_028693031.1 | 1937 | 72 | |||
| XM_028837199.1 | XP_028693032.1 | 2750 | 72 | |||
| XM_028837201.1 | XP_028693034.1 | 2163 | 72 | |||
| XM_028837202.1 | XP_028693035.1 | 2154 | 72 | |||
| XM_028837203.1 | XP_028693036.1 | 1820 | 72 | |||
| XM_028837204.1 | XP_028693037.1 | 2163 | 72 | |||
| XM_028837207.1 | XP_028693040.1 | 1820 | 72 | |||
| XM_028837216.1 | XP_028693049.1 | 1976 | 72 | |||
| GenBank mRNA | JU325659.1 | - | 1607 | 72 | ||
| JU325661.1 | - | 1607 | 72 | |||
| JU325662.1 | - | 1607 | 72 | |||
| JU476884.1 | - | 1844 | 72 | |||
| JV636327.1 | - | 1844 | 72 | |||
More Information
Set Membership: |
|

PHACTR4
Genomic Map