Assay Details
| Gene Symbol: RCCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193932.2 | NP_001180861.2 | 982 | 59 | ||
| GenBank mRNA | JU325061.1 | - | 839 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RCCD1| Gene Symbol: RCCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193932.2 | NP_001180861.2 | 982 | 59 | ||
| GenBank mRNA | JU325061.1 | - | 839 | 59 | ||
Set Membership: |
|