Assay Details
| Gene Symbol: SIGLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838835.1 | XP_028694668.1 | 200 | 88 | ||
| XM_028838836.1 | XP_028694669.1 | 201 | 88 | |||
| Gene Symbol: SIGLEC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838837.1 | XP_028694670.1 | 255 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028838835.1, XM_028838836.1 transcripts.
Set Membership: |
|

SIGLEC11
Genomic Map