Assay Details
| Gene Symbol: RFFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119059.2 | XP_014974545.1 | 788 | 63 | ||
| XM_015119062.2 | XP_014974548.1 | 957 | 63 | |||
| XM_015119063.2 | XP_014974549.1 | 809 | 63 | |||
| XM_015119065.2 | XP_014974551.1 | 953 | 63 | |||
| XM_015119066.2 | XP_014974552.1 | 793 | 63 | |||
| XM_015119067.2 | XP_014974553.1 | 809 | 63 | |||
| XM_015119071.2 | XP_014974557.1 | 938 | 63 | |||
More Information
Set Membership: |
|

RFFL
Genomic Map