Assay Details
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194627.2 | NP_001181556.2 | 808 | 69 | ||
XM_015135944.2 | XP_014991430.1 | 2386 | 69 |

More Information
Set Membership: |
![]() ![]() |
