Assay Details
| Gene Symbol: PAIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257252.1 | NP_001244181.1 | 351 | 132 | ||
| XM_015140811.2 | XP_014996297.1 | 528 | 132 | |||
| GenBank mRNA | CN642792.1 | - | 450 | 132 | ||
| JU334834.1 | - | 369 | 132 | |||
| JV047268.1 | - | 447 | 132 | |||
| JV566125.1 | - | 490 | 132 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PAIP2
Genomic Map