Assay Details
| Gene Symbol: RHOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015153008.2 | XP_015008494.1 | 1105 | 64 | ||
| XM_028830010.1 | XP_028685843.1 | 1708 | 64 | |||
| GenBank mRNA | JU471149.1 | - | 505 | 64 | ||
| JU488486.1 | - | 583 | 64 | |||
More Information
Set Membership: |
|

RHOF
Genomic Map