Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140679.2 | XP_014996165.1 | 615 | 89 | ||
| XM_015140680.2 | XP_014996166.1 | 1168 | 89 | |||
| XM_015140684.2 | XP_014996170.1 | 614 | 89 | |||
| XM_015140686.2 | XP_014996172.1 | 606 | 89 | |||
| XM_028833258.1 | XP_028689091.1 | 603 | 89 | |||
| XM_028833275.1 | XP_028689108.1 | 551 | 89 | |||
| GenBank mRNA | JV051069.1 | - | 577 | 89 | ||
More Information
Set Membership: |
|

OMA1
Genomic Map