Assay Details
| Gene Symbol: ZBTB49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096571.4 | XP_001096571.2 | 1429 | 86 | ||
| XM_028848345.1 | XP_028704178.1 | 1429 | 86 | |||
| XM_028848346.1 | XP_028704179.1 | 2278 | 86 | |||
| GenBank mRNA | JU474521.1 | - | 1331 | 86 | ||
| JV644022.1 | - | 1359 | 86 | |||
More Information
Set Membership: |
|

ZBTB49
Genomic Map