Assay Details
| Gene Symbol: REEP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140593.2 | XP_014996079.1 | 524 | 89 | ||
| GenBank mRNA | JU319599.1 | - | 327 | 89 | ||
| JU473232.1 | - | 329 | 89 | |||
| JV044507.1 | - | 308 | 89 | |||
| JV723172.1 | - | 300 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

REEP5
Genomic Map