Assay Details
| Gene Symbol: C1GALT1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260840.2 | NP_001247769.1 | 125 | 90 | ||
| GenBank mRNA | JU334152.1 | - | 137 | 90 | ||
| JU475280.1 | - | 156 | 90 | |||
| JV046840.1 | - | 127 | 90 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260840.2 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C1GALT1C1
Genomic Map