Assay Details
| Gene Symbol: TAMM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193810.2 | NP_001180739.2 | 277 | 96 | ||
| XM_028843106.1 | XP_028698939.1 | 412 | 96 | |||
| XM_028843107.1 | XP_028698940.1 | 412 | 96 | |||
| XM_028843108.1 | XP_028698941.1 | 396 | 96 | |||
| GenBank mRNA | JV051924.1 | - | 147 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028843108.1 transcripts.
Set Membership: |
|

TAMM41
Genomic Map